Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Advansta WesternBright™ ECL HRP Substrate Kits
Supplier: Advansta
K-12045-C20
490005-016EA 86.4 USD InStock
490005-016
Advansta WesternBright™ ECL HRP Substrate Kits
Electrophoresis Reagents Western Blotting Reagents
WesternBright ECL is a horseradish peroxidase substrate optimized for chemiluminescent Western blots imaged using x-ray film. WesternBright ECL requires as little as ten times less antibody than other substrates, allowing you to save precious antibodies and samples. Additionally, the WesternBright ECL signal is long lasting, allowing multiple exposures to be carried out without substantial signal decay.
  • Shorter Exposures- Stronger signal than Amersham™ ECL or SuperSignal® West Pico
  • Sensitive- Detect Low pg Protein Amounts
  • Save antibody- Use up to Ten-Times Less Antibody
  • Long-lasting signal- Image Blots up to Hours After Substrate Incubation, No Rush to Get to Dark Room
  • Room temperature stable- One Year Shelf Life at Room Temperature.
The best choice for routine, film-imaged Western blots. WesternBright ECL is more sensitive than other chemiluminescent substrates, so you can conduct shorter exposures, load less sample, or use up to ten-times less antibody. Save time and money with WesternBright ECL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty