Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tetra-Chloroauric(III) acid trihydrate ACS

Supplier: VWR
Danger
Synonyms: Hydrogen tetrachloroaurate (III) trihydratetetra-Chloroauric acid trihydrateGold Chloride hydrateHydrogen tetrachloroaurate(III) trihydrateGold(III) chloride trihydrate

VWRV0240-5G
490005-234CS 27491.38 USD InStock
490005-234
tetra-Chloroauric(III) acid trihydrate ACS
tetra-Chloroauric (III) acid trihydrate
Insoluble in Ether 0.1%. Purity (Au) 49.0%

Certifications: ACS Grade: Materials conforming with the specifications and procedures outlined in American Chemical Society Specifications, 9th Edition, 2000.


Formula: HAuCl₄·3H₂O
MW: 393.83 g/mol
Melting Pt: 30 °C
Density: 3.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149904
CAS Number: 16961-25-4
EINECS: 240-948-4
UN: 1759
ADR: 8,III
Order Now
Specification Test Results
Alkalies & Other Metals 0.2%
Insoluble in Ether = 0.1%
Purity (Au) = 49%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty