Would you like to log in again?
SP Bel-Art Vikem® Vinyl-coated Lead Ring, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18307-0010 F18307-0020 F18307-0015
490005-576EA 64.8 USD InStock
490005-576 490005-574 490005-572
SP Bel-Art Vikem® Vinyl-coated Lead Ring, Bel-Art Products, a part of SP
Weights Flask Weights
Vikem Vinyl-Coated Lead Rings are available in a range of sizes to fit Erlenmeyer-style flasks.
  • Vinyl coating
  • Available in a range of sizes
  • Chemically resistant
  • Non-scratching
Reduces breakage, floating, and spillage.
The non-slip Vikem surface seats securely around the neck of the flask. Use lead rings to stabliize labware in water baths. This weight prevents floating and reduces breakage and spilling. The cushioned surface is chemically resistant and willl not scratch glass labware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty