Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
SP Bel-Art Spinbar® Color Octagon Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art
Spinbar®
F37109-0012 F37109-0009 F37109-0030 F37109-0008 F37109-0001 F37109-0028 F37109-0011 F37109-0029 F37109-0020 F37109-0019 F37109-0036 F37109-0006 F37109-0003 F37109-0034 F37109-0021 F37109-0031 F37109-0032 F37109-0010 F37109-0033 F37109-0004
74950-246EA 13.48 USD InStock
74950-246 74950-258 490005-588 74950-244 490005-586 74950-242 74950-254 74950-240 58948-030 76001-824 47750-128 47746-177 76001-822 76001-820 76001-886 76001-882 47750-122 47750-124 74950-248 74950-238
SP Bel-Art Spinbar® Color Octagon Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
  • PTFE low-friction coating
  • Durable and inert
  • Interrupted profile
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
Interrupted profile provides greater surface area and added turbulence
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Choose from red, yellow or blue.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty