Would you like to log in again?
Proteinase K
Supplier: Meridian Bioscience
BIO-37039 BIO-37037
490005-956EA 222.7 USD InStock
490005-956 490005-958
Proteinase K
Enzymes
Proteinase K is an enzyme used throughout Molecular Biology as a protein denaturant
Molecular Biology grade Proteinase K is DNAse and RNAse free and is used for inactivation of RNAses and DNAses during nucleic acid extraction and protein modification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty