Would you like to log in again?
DNA Loading Buffer 5X, Bioline
Supplier: Bioline
BIO-37045
490005-960EA 83.4 USD InStock
490005-960
DNA Loading Buffer 5X, Bioline
Buffers Electrophoresis Buffers Loading Buffers
DNA loading buffer blue (5X) is a convenient ready-to-use solution to prepare samples for DNA electrophoresis.
  • Ready-to-use
  • Dye visible in gel
  • Guaranteed batch-to-batch reproducibility
DNA loading buffer blue is premixed with bromophenol blue and agarose gel which allows to monitor DNA migration and therefore increase the versatility of your DNA analysis. This can be used for monitoring migration rate during agarose electrophoresis and in loading DNA samples onto agarose gels.
Caution: Repeated freeze/thaw cycles should be avoided.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty