Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mini Tube Rotator, Boekel Scientific

Supplier: Boekel Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

260750
490006-048EA 626 USD InStock
490006-048
Mini Tube Rotator, Boekel Scientific
Shakers and Mixers Rotating/Rocking Shakers
The variable speed Mini Tube Rotator includes an LCD display and has a small footprint for your bench.

  • Variable speed
  • Adjustable mixing angle
  • Cold room and incubator compatible
  • LCD display

This rotator takes up minimal space on the laboratory bench while still providing a complete 360° mixing rotation and a 0 to 90° mixing angle.

Certifications: AC Adapter is UL, CSA and CE approved.

Ordering information: Interchangeable carousels for 1.5/2.0, 10/15 and 50 ml tubes are available.
Supplied with carousel (76326-396) to hold 36×1.5/2.0 ml tubes.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty