Would you like to log in again?
Dilux™ Dilution Reservoir
Supplier: Dilux
D-1003 D-1002
490006-654EA 128.92 USD InStock
490006-654 490006-656
Dilux™ Dilution Reservoir
Reagent Reservoirs
Designed for making serial dilutions of biological samples and chemical reagents using multichannel pipettors, the 12-chamber reservoir holds up to 5 mL of sample
The rounded sides and bottom of the chambers permit complete sample withdrawal. A numerical code inscribed for each chamber ensures accurate sample reference. The fitted lid minimizes sample evaporation and contamination during incubation or storage, while the broad base of the container ensures bench stability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty