Would you like to log in again?
Diversified Biotech Biodegradable Reagent Reservoirs
Supplier: Diversified Biotech
REBP-5000 REBP-2000 REBP-1000 REBP-3000
490006-884CS 244.68 USD InStock
490006-884 490006-888 490006-886 490006-890
Diversified Biotech Biodegradable Reagent Reservoirs
Reagent Reservoirs
Disposable, Sterile, and Safe for the Environment.
  • Biodegradable- Using a special additive to PET, our reservoirs will safely degrade over time
  • Packaging 100% Eco-Friendly- bags are biodegradable & cardboard cases are recyclable
  • Unique- "Trough within a trough" maximizes reagent recovery
  • Pour-off spouts on all four corners
  • Stable- A wide base and reinforced sidewalls avoids spills and provides stability
  • Graduated marks on inside wall
  • Sterile- Sterilized by gamma irradiation
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty