Would you like to log in again?
Angled Bottle Rest, Diversified Biotech
Supplier: Diversified Biotech
REST-1000
490006-914EA 161.82 USD InStock
490006-914
Angled Bottle Rest, Diversified Biotech
Support Stands
Supports round and square bottles for pipetting Comfort
  • Durable- Made of long-lasting acrylic and polypropylene
  • Ideally sized- Accommodates 50 mL to 1 L square or round bottles
  • Space saving- Compact design takes up little benchtop space
Pipette at a comfortable 45˚.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty