Would you like to log in again?
Polypropylene Graduated Cylinders, Dynalon
Supplier: Dynalon
239055 239075
490007-178CS 391.59 USD InStock
490007-178 490007-176
Polypropylene Graduated Cylinders, Dynalon
Cylinders Measuring Cylinders
This graduated cylinder is molded of chemically resistant, translucent polypropylene.
  • Stable, hexagonal bases with non-slip anti-suction feature
  • Very easy to read with virtually no meniscus
Graduations are permanently molded on cylinders and will not fade after repeated autoclaving. Meets stringent international quality criteria according to ISO 9000 standards. Tolerances according to DIN 12 680TT, Class B.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty