Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Dynalon Bio-bin® Waste Disposal Containers
Supplier: Dynalon
Bio-bin®
797303-6300 797303-0002 797303-0030 797303-0006 797303-6200
490007-232EA 129.64 USD InStock
490007-232 490007-226 490015-310 490007-228 490007-230
Dynalon Bio-bin® Waste Disposal Containers
Boxes Waste Disposal Boxes
Ideal use in academic, clinical, government and industrial laboratories for a wide rang of non-sharps biomedical and laboratory waste that require special heat treatment such as autoclaving or incineration before disposal.
  • Space efficient, lightweight fold-flat design
  • Ergonomic design, built-in lid and handles
  • Stackable post-use
  • Temporary & permanent seals
  • Printed user instructions & user ID/date information on lid
  • Waterproof construction
  • Built-in super absorbent Bio-matt to soak up any excess liquid
  • Autoclavable
6 liter Bio-bin is ideal for use with Biological Safety Cabinets.
Certifications: UN 3291 approved for handling Regulated Bio-Medical Waste.
Delivery information: Supplied with 2" x 2" Biohazard Waste Labels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty