Would you like to log in again?
SealPlate®, SealPlate® MiniStrips™, and ThinSeal™ Plate Sealing Film, Excel Scientific
Supplier: Excel Scientific
100-SEAL-PLT SPS-2X8-50 STR-SEAL-PLT STR-THIN-PLT 100-THIN-PLT SP-2X8-50
490007-366EA 86.65 USD InStock
490007-366 490007-344 490007-312 490007-308 490007-360 490007-362
SealPlate®, SealPlate® MiniStrips™, and ThinSeal™ Plate Sealing Film, Excel Scientific
Seals Microplate Seals
SealPlate® 50 µm thickness polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover.
SealPlate® MiniStripsTM sealing films are identical to SealPlate films above, but designed for sealing only one or two 8-well rows at a time on either strip-well or standard plates whenever rows must be selectively protected or accessed. Each MiniStrips film is 19.0 x 103.2 mm, including two positioning tabs. The long dimension with the tabs removed is 82.6 mm. MiniStrips are supplied in sheets of four from which strips can be peeled individually or two-at-a-time for application to plates. Sterile product is packaged in 100 µm tamper-evident bags of 100. Two tabs on each strip for easy application and removal. Perforated tabs can be removed or sealed around plate edge. Can be used as secondary seal to cover sealing films that have been pierced. More convenient.
ThinSeal™ polyester sealing films are just 25 µm thick, clearer and marginally pierceable with single-channel pipettes for sample recovery. ThinSeal sealing film dimensions are 79.4 x 141.0 mm. Functional temperature range from -40°C to 120°C. Available sterile and nonsterile
Secure sealing of all wells eliminates "edge effects" in sensitive ELISA assays. SealPlate sealing films can also be used with tissue-culture plates for short-term storage, incubation, and containment of biohazards. SealPlate sealing films are non-pierceable. Each SealPlate sealing film measures 79.4 x 146.1 mm. Functional temperature range from -40°C to 120°C. Available sterile and nonsterile.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty