Would you like to log in again?
ThermalSeal® MiniStrips™ Seals, Excel Scientific
Supplier: Excel Scientific
TS-2X8-50
490007-372EA 54.2 USD InStock
490007-372
ThermalSeal® MiniStrips™ Seals, Excel Scientific
Seals Microplate Seals
ThermalSeal MiniStrips sealing films are identical to ThermalSeal sealing films, but designed for sealing only one or two 8-well rows at a time on PCR plates or strip tubes.
  • Two tabs on each strip for easy application and removal
  • Perforated tabs can be removed or sealed around plate edge
  • Heated resistant, recommended for temperatures from -40°C to 120°C
  • Certified DNase-, RNase-, and nucleic-acid-free
They are now supplied in convenient sheets of four. Each sealing film strip is 19.0 x 97.3 mm, including two removable positioning tabs. The long dimension with tabs removed is 76.2 mm.
Packaging: Each package of 200 contains 50 sheets of four sealing film strips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty