Would you like to log in again?
NitroPlus Transfer Membranes, GVS Filter Technology
Supplier: GVS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NitroPlus
1212632 1212592 1213141 1212604 1212595 1212602 1212721 1212669 1212596 1212601 1212591 1212597 1212590 1212690 1214560
490007-430PK 546.77 USD InStock
490007-430 490007-420 490007-454 490007-432 490007-422 490007-444 490007-434 490007-424 490007-446 490007-436 490007-448 490007-426 490007-428 490007-418 490015-798
NitroPlus Transfer Membranes, GVS Filter Technology
Transfer Membranes
Supported Nitrocellulose (MCE) membranes.
  • Supported for rigorous handling and multiple reprobings
  • Strong construction will not curl, bend, or crack after baking
  • High sensitivity, low background
  • BSA binding capacity up to 100 ug/cm2
  • Triton-free
GVS Filter Technology NitroPlus Supported Transfer Membranes combine the binding characteristics of a nitrocellulose membrane with the strength of a nylon membrane. NitroPure can easily be used in any protocol utilizing unsupported nitrocellulose transfer membranes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty