Would you like to log in again?
Dual Solutions™ Reservoirs
Supplier: Heathrow Scientific
Dual Solutions™
HS20821A
490007-612PK 150.7 USD InStock
490007-612
Dual Solutions™ Reservoirs
Reagent Reservoirs
Multi-functional reagent reservoir has a single compartment 50 mL basin on one side and 12 slotted 5 ml basins on the other side.
  • Angled wells guide pipette tips toward a center position to help avoid cross contamination and waste
  • Individual 5 mL basin wells numerical imprinted to identify samples
  • Cover included to prevent evaporation and contamination
  • Made of polypropylene
Dimensions: 12.5×5×3.7 cm (4.9×2×1.5").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty