Would you like to log in again?
Storage Racks, Microtube Racks 80-Wells
Supplier: Heathrow Scientific
HS23263B HS23263A
490007-622EA 207.3 USD InStock
490007-622 490007-624
Storage Racks, Microtube Racks 80-Wells
Racks Tube Racks
These racks helps to keep large numbers of microtubes organized on benchtop.
  • Strong, clear acrylic construction
  • Plenty of room to slide the racks in and out
File system accommodates up to eight 80-well microtube racks with either 1.5 or 2.0 ml microtubes in the racks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty