Would you like to log in again?
Shelton Scientific BETA-GARD™ Storage Containers, IBI Scientific
Supplier: IBI Scientific
BETA-GARD™
RB-187
490007-710EA 395.86 USD InStock
490007-710
Shelton Scientific BETA-GARD™ Storage Containers, IBI Scientific
Boxes Beta/Gamma Radiation Boxes
BETA-GARD Storage Containers provide safe storage and transport of beta radition emitting samples.
The storage containers hold most test tube racks, small bottles, and vials. Constructed of heavy-duty 1/2" cast acrylic for maximum radiation protection and is refrigerator/freezer compatible.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty