Would you like to log in again?
GLOVE LATEX PF EGRIP LG 100BX
Supplier: Ansell Healthcare
Glove^ General purpose and examination glove, Disposable, High Five*, Grade: Medical, Material: Latex, Powder free, Color: Off-White, Size: Large, Surface Finish: Rough, Smooth, Cuff Type: Beaded.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE LATEX PF EGRIP LG 100BX 490008-144 L973 Box of 100 Retrieving
Case of 1000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty