Would you like to log in again?
MBP Snap-Cap Centrifuge Tubes
Supplier: Molecular BioProducts
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3446 3448 3434
490008-460CS 426.66 USD InStock
490008-460 490008-464 490008-462
MBP Snap-Cap Centrifuge Tubes
Tubes Microcentrifuge Tubes
All tubes are certified RNase/DNase-, RNA/DNA-, ATP-, and pyrogen-free, and we believe that there are no tubes better suited for the everyday rigors of RNA work. PurePak bag is designed to protect the integrity of the tubes.
  • 0.6 mL tubes, rated 30,000 x g
  • 1.5 mL tubes, rated 26,000 x g
  • 2.0 mL tubes, rated 25,000 x g
  • 4 mL puncture-proof bag
  • Zip closure for easy resealing
  • 10 individually sealed bags of tubes help protect against contamination
  • Graduated Microcentrifuge Tubes with Frosted Flat Cap and Side
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty