Would you like to log in again?
Labnet ENDURO™ Horizontal Gel Boxes, Labnet International
Supplier: Labnet
E1010-10 E1015-10 E1007-10 E1015-15 E1020-20 E1007-7
490008-958EA 884.7 USD InStock
490008-958 490008-956 490008-966 490008-964 490008-962 490008-960
Labnet ENDURO™ Horizontal Gel Boxes, Labnet International
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Enduro Gel Boxes have been designed with safety and ease of use in mind, molded from UV transparent acrylic, for leak-proof performance. Cassettes enclose the electrodes to protect them from breakage and allow for easy cleaning and replacement. Positioning slots on the sides of the trays hold combs in place while pouring gels. The supplied combs are 1.5 mm thick and are adjustable to control well depth.
  • Rubber gates for easy, tapeless gel casting
  • Adjustable height combs
  • Loading guides aid in well visualization
  • UV transparent gel trays
  • Quality construction
  • 1-year warranty
Each gel tray is supplied with a guide that can be placed underneath to aid in visualizing wells when loading. The domed lid and drip ring prevent condensation from falling onto the gel. The corrosion-resistant platinum electrodes are located in cassettes to protect them during cleaning and provide for easy replacement. The lids fit on the gel boxes in only one direction. A drip ring on the lid recycles condensed buffer and prevents it from dripping directly onto the gel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty