Would you like to log in again?
Labnet Excel™ Electronic Pipettes, Labnet International
Supplier: Labnet International
P3600L-1200 P3608L-10 P3608L-1200 P3600L-20 P3612L-1200 P3608L-200 P3612L-20 P3600L-10 P3600L-200
490009-216EA 1833.72 USD InStock
490009-216 490009-228 490009-218 490009-212 490009-224 490009-214 490009-226 490009-230 490009-222
Labnet Excel™ Electronic Pipettes, Labnet International
Pipettes Micropipettes
The Excel™ electronic pipettes are multifunctional units with operational modes for standard and reverse pipetting as well as multiple and sequential pipetting, mixing of samples and serial dilutions.
  • Exceptional accuracy and precision
  • Simple, precise volume setting
  • Fully motorized piston drive
  • Multifunctional with six operating modes
  • Virtually eliminates pipetting force
  • User friendly operator interface
  • Rechargeable Lithium-ion battery
  • Automatic calibration using external weight
All functions are easily programmed with one hand on the well positioned display and keypad. A fully motorized piston drive and auto-calibration guarantee excellent accuracy and reproducibility. A removable, rechargeable Lithium-ion battery ensures many hours of performance between charges.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty