Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Disruptor Genie® Cell Disruptor Homogenizer, Scientific Industries

Supplier: Scientific Industries
SI-BG01 SI-DD38 SI-0565 SI-BG05 SI-BG05 SI-D238 SI-BG01 SI-DD48 SI-D248
89426-874EA 51.39 USD InStock
89426-874 89202-318 490010-032 89202-320 89202-324 490010-034 89202-314 89426-876 89202-316
Disruptor Genie® Cell Disruptor Homogenizer, Scientific Industries
Homogenizers Benchtop Homogenizers
Economical alternative to ultransonic disruptors.

  • Rapid hands-free disruption of up to 12 microtubes (1.5 mL or 2.0 mL) at a time
  • Use with Disruptor Beads, made from spherical, lead free, soda lime glass
  • Suitable for use in cold rooms or incubators
  • Supplied with Microtube holder for 12 microtubes (tubes not included), and Pop-Off Cup
  • Adjustable digital speed alarm
  • 2-year warranty included

Dramatically increase cell disruption efficiency

This Disruptor is available in analog or digital models, allows for Hands-Free disruption of up to twelve 1.5 or 2.0ml microtubes with timer from 0-15 minutes or continuous for analog models, and from 0-99 minutes or continuous for Digital models. Provides multi-directional action, which simultaneously agitates and vortexes at high speed, dramatically increasing cell disruption or sample resuspension efficiency. Suitable for use in cold rooms or incubators. Ideal for difficult glass bead procedures - cell disruption/homogenization of yeast, bacteria, plant and animal tissue or anything that requires extremely violent high speed agitation. Digital model incorporates an adjustable speed alarm. Remove microtube holder and attach the supplied Pop-off Cup for high speed vortexing of single test tubes.

The patented Disruptor Genie provides multi-directional action, which simultaneously agitates and vortexes at high speed, dramatically increasing cell disruption or sample resuspension effeciency.

Certifications: ETL listed (120V model). CE marked.

Accessories information: Cell disruption is optimized with the use of beads (not included with product).

Ordering information: Universal Microtube Holder and Pop-off Cup included. The 230V model is supplied without a plug.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty