Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Cryostore® Storage Racks, Simport
Supplier: Simport
T314-581R T314-281G T314-281Y T314-581B T314-2100Y T314-581G T314-281B T314-281R T314-2100R T314-2100G T314-2100B
490010-408CS 265.81 USD InStock
490010-408 490010-392 490010-406 490010-390 490010-396 490010-394 490010-410 490010-388 490010-400 490010-398 490010-402
Cryostore® Storage Racks, Simport
Racks Cryogenic Vial Racks
Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -190°C and 121°C and are autoclavable
A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Being printed with a series of squares (numbered from 1 to 100), the surface accepts writing markers for better inventory control. A unique color coding system uses colored plastic grids inserted to separate the cover from the base of the 81-place boxes. Boxes made to accept 100 tubes have a colored base instead of a grid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty