Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sonics Accessories for 50-Watt Liquid & 130-Watt Ultrasonic Processors
Supplier: Sonics
830-00118 630-0423 630-0422 630-0435 630-0451 630-0561
490010-576EA 142.64 USD InStock
490010-576 490010-554 490010-562 490010-568 490010-566 490010-556
Sonics Accessories for 50-Watt Liquid & 130-Watt Ultrasonic Processors
Homogenizers Benchtop Homogenizers
Optional Probes
Probes with smaller diameter produce greater intensity of cavitation, but the energy released is restricted to a more concentrated field. Probes with a larger tip diameter produce less intensity, but can process larger samples.Footswitch is ideal for hands-free operation. Includes a 10-ft cable. Use with S-2034-130 only.Converter Clamp supports a 32 mm (1.25") converter and fits a 13 mm (0.5") rod. Chemical-resistant, reinforced plastic.Sound-abating Enclosure permits extended processing without discomfort by reducing noise to a comfortable level. Support rod and converter clamp are included. Measures 12" W x 20" H x 12" D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty