Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Handheld Refractometer, 0 to 100 ppt Salinity, ATC, STX-3

Supplier: Heathrow Scientific
43036
490011-000EA 204.52 USD InStock
490011-000
Handheld Refractometer, 0 to 100 ppt Salinity, ATC, STX-3
Refractometers Handheld Refractometers
The handheld salinity refractometer features the largest, easiest-to-read scale available with clear, sharp figures, screen shading and incredible contrast. This significantly enhances overall precision, even when several different people use the refractometer.

  • Quick results: High quality optical glass prism and all-metal housing design allows for quick temperature equilibrium
  • Accurate and sharp readings: Large scale is best in class delivering clear, sharp figures, screen shading and incredible contrast for overall accuracy
  • Protective grip: The rubber grip reduces hand slipping and guards against user’s hand heat to maintain accuracy
  • Automatic Temperature Compensation (10 to 30˚C/50 to 86 °F)
  • Scales: Salinity/Specific Gravity of Seawater
  • Length: 185 mm

The handheld refractometer is exceptionally simple to operate and provides quick, accurate measurements for the concentration of nearly any aqueous solution in a wide range of applications. Made durable with industrial-grade design features to provide years of reliable use even in the field. Rugged metal housing construction with slip-resistant rubber grip and a non-roll stand to protect from rolling off surfaces.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty