Would you like to log in again?
Serological Pipettes
Supplier: Vee Gee Scientific
2011A-10 2011A-25 2011A-5
490011-032CS 287.11 USD InStock
490011-032 490011-034 490011-036
Serological Pipettes
Pipettes Serological Pipettes
Serological pipettes are made out of high-quality borosilicate glass. They also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges.
  • Wide range of sizes to choose from
  • Reusable
  • Calibrated to deliver
Serological pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of liquid handling needs.
Certifications: Meets ASTM E-1044 Style 1, Class A specifications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty