Would you like to log in again?
100 ml Volumetric Pipette, Class A
Supplier: Vee Gee Scientific
2040A-100
490011-056CS 576.14 USD InStock
490011-056
100 ml Volumetric Pipette, Class A
Pipettes Graduated Pipettes
These volumetric pipettes are graduated to the tip and made out of high-quality borosilicate glass. These pipettes also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges.
  • Made out of high-quality borosilicate glass
  • Reusable
  • Color-coded with permanent enamel markings
  • Calibrated To Deliver
Certifications: Meets ASTM E-969 Class A Specifications and USP Standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty