Would you like to log in again?
Vee Gee Volumetric Flasks with Glass Stopper
Supplier: Vee Gee Scientific
2306A-2000 2306A-50PK 2306A-1000PK
490011-060CS 468.77 USD InStock
490011-060 490011-062 490011-058
Vee Gee Volumetric Flasks with Glass Stopper
Flasks Volumetric Flasks
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers. Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 68 °F (20 °C)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Wide range of sizes available
Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.
Certifications: Meets ASTM E-288 Class A specifications & USP standards for glassware
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty