Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vee Gee Graduated Cylinders, Class B

Supplier: Heathrow Scientific
Vee Gee
2351-250 2351-1000 2351-500 2351-2000 2351-50 2351-100 2351-10 2351-25
76546-172CS 2122.86 USD InStock
76546-172 76546-170 76546-168 76546-178 490011-068 76546-166 76546-176 76546-174
Vee Gee Graduated Cylinders, Class B
Cylinders Measuring Cylinders
These high-quality graduated cylinders are made of premium borosilicate glass and comply with ASTM E-1272 Class B standards, guaranteeing your investment in high-quality glassware.

  • Made out of high-quality borosilicate glass
  • Double metric scale
  • Graduated 'to contain'
  • Contains durable white enamel graduation markings
  • Wide range of sizes available (10 to 2000 ml)
  • Feature hexagonal bases for added stability
  • All sizes (except 2000 ml) come with a protective bumper guard
  • Calibration temperature: 20 °C (68 °F)

With their exceptional quality, precise accuracy, and ease of use, they are a great tool to use for all of your liquid handling needs.

Each graduated cylinder features a double metric scale, which is calibrated to contain, ensuring that the results of your experiments are precise and trustworthy. These graduated cylinders also feature an easy-pour spout, which allows for a controlled and drip-free pour, preventing spillage. All sizes come with a hexagonal base for added stability and contain white enamel graduation markings that are easy to read, minimising measurement errors.

Certifications: Exceeds ASTM E-1272 Class B standards.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty