Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art Holdfast Graduated Cylinders, Bel-Art Products, a part of SP

Supplier: Bel-Art

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

F28461-1000
490012-188EA 47.55 USD InStock
490012-188
SP Bel-Art Holdfast Graduated Cylinders, Bel-Art Products, a part of SP
Cylinders Measuring Cylinders
Reduce fatigue when repetitive measuring and pouring is required.

  • Graduated markings
  • Easy to read for measuring
  • Convenient handle for pouring
  • Steam autoclavable at 121°C (250°F)

The large handle provides a secure grip for safer, more controlled and convenient pouring. A double scale is accurate and easy to read. The wide hexagonal base prevents tip over and rolling on the benchtop. Hand opening: 89 x 29 mm (3 1/2 x 1 1/8").

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty