Would you like to log in again?
RACK RND FLOATING 1000ML 4/PK
Supplier: Bel-Art Products, a Part of SP
Rack^ Economy round floating rack, 20 places, Asnug fit retains the tubes that can be easily released by lightly pressing the rack against the bench until the legs touch, Polypropylene racks float maintaining 1.5ml tubes immersed the in bath with tops held above the water level
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK RND FLOATING 1000ML 4/PK 490012-416 F18875-6411 Case of 4 Retrieving
Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty