Would you like to log in again?
Sample Vials with Hinged Lid, Dynalon
Supplier: Dynalon
226245-25 226245-10 226245-50
490012-420CS 1443.27 USD InStock
490012-420 71070-206 71070-214
Sample Vials with Hinged Lid, Dynalon
Vials
General-purpose containers feature push-in friction fit hinged lids.
Lightweight vials have a wall thickness that is approximately 1 mm. LDPE construction withstands temperatures up to 80 °C (176 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty