Would you like to log in again?
Vinyl Tubing, Dynalon
Supplier: Dynalon
610855-09
490012-467CS 85.92 USD InStock
490012-467
Vinyl Tubing, Dynalon
Tubing Transfer Tubing
Vinyl food grade tubing that resists fruit acids, lye, alcohol, greases, and oils.
  • Non-toxic
  • Inert
  • Flexible
  • Transparent
  • Smooth
  • Chemical resistant
Vinyl Food Grade Tubing is designed for use in the laboratory. Offering glass-like transparency, the smooth, dense bore will not harbor bacteria or provide a surface receptive to sediment build-up. Conforms to all FDA, USDA, and 3-A milk handling standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty