Would you like to log in again?
SLIDE MICROSCOPE SUPERFROST 25X75MM PK72
Supplier: Erie Scientific
Slide^ Superfrost* Plus Micro Slide Opaque coating retains most laboratory marking methods. Specially treated for frozen tissue sections and cytology preparation applications. Improved tissue adherence during microwave staining. Thickness: 1mm. Size: 25Lx7
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SLIDE MICROSCOPE SUPERFROST 25X75MM PK72 490013-246 4951PLUS-001 Case of 1440 Retrieving
Pack of 72 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty