Would you like to log in again?
MBP Locking-Cap Microcentrifuge Tubes, Graduated
Supplier: Molecular BioProducts
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
L-508-GRDS L-510-GRDS L-502-GRD L-502-GRD-MIXED L-508-GRD-MIXED L-508-GRD L-510-GRD-Y L-510-GRD-R
490016-249BG 68.2 USD InStock
490016-249 490016-247 490016-245 490016-243 490014-117 490016-241 490016-251 490014-119
MBP Locking-Cap Microcentrifuge Tubes, Graduated
Tubes Microcentrifuge Tubes
A superior lock for safe and convenient centrifuging, boiling, freezing, incubation or storage of samples.
  • Smooth surfaces reduce the possibility of ripped or punctured gloves
  • Manufactured of ultra clear, autoclavable polypropylene
Locking microcentrifuge tubes offer the same sample security as found with screw cap tubes yet maintain the same ease of use as a snap cap. Unique design allows visual confirmation that the tube is locked. The pierceable cap snaps closed and the lock slips easily into place with a simple one hand operation. The larger, rounder cap design proves less stressful to the thumb.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty