Would you like to log in again?
CappAero™ Premium Multi-channel Pipettes
Supplier: CAPP
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C300-12 C10-12 C50-8 C50-12 C10-8 C300-8 C200-12 C200-8
490014-550EA 1904.55 USD InStock
490014-550 490005-280 490014-552 490005-272 490005-276 490005-264 490014-548 490005-274
CappAero™ Premium Multi-channel Pipettes
Pipettes Micropipettes
Engineered for maximum performance and comfort.
  • Light weight great ergonomic fit in hand
  • Low plunger forces
  • Fully autoclavable
Double O-ring seal on shaft for easy mounting and ejection of the tips. Stainless steep piston for excellent accuracy and precision. Virtually no maintenance required- No annual seals and O-rings to replace. Aluminum alloy shaft is virtually unbreakable- No more shaft replacements. Interchangeable control knobs for rapid volume setting changes. Calibrate your Capp Pipettes with no down time. 3-year warranty. The 30 to 300 µl, variable from 125-3 µl only. Below 125 µl please use fixed volume controllers. CappAero is a trademark of Capp A/S.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty