Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Advansta AdvanStain Scarlet™ Staining Kits
Supplier: Advansta
K-11072-250 K-11072-B50 K-11072-C25
490016-810EA 45.32 USD InStock
490016-810 490015-240 490015-238
Advansta AdvanStain Scarlet™ Staining Kits
Stains Microbiology Stains
  • Detect less than a nanogram of protein per spot
  • Simple 3-hour protocol
  • Stain gels or membranes
  • Destain for downstream western blotting or mass spectrometry
  • Clean background with no speckling for better data
  • Biodegradable; contains no heavy metals
  • Increased safety and simple disposal
  • Image with any laser or CCD imaging system
AdvanStain Scarlet is a fluorescent stain for gels and blots that allows sensitive and quantitative visualization of proteins. Gels and blots stained with AdvanStain Scarlet can be imaged on any fluorescent imaging system, including laser- and CCD-based systems, with a detection limit of less than 1 ng of protein per band or spot. Fully compatible with downstream Western blotting or mass spectrometry, AdvanStain Scarlet is non-toxic and biodegradable, for safe and simple disposal. Compatible excitation wavelengths include green (543, 532 nm), blue (488 nm), violet (405 nm) and UV (302, 365 nm). Maximum emission wavelength is 610 nm.
AdvanStain Scarlet outperforms both Coomassie blue and SYPRO® Ruby in time and sensitivity. Gels were stained with AdvanStain Scarlet, SYPRO Ruby, or Coomassie blue stain, according to the manufacturers' protocols and imaged on a CCD imager. AdvanStain Scarlet is sensitive, quantitative, non-toxic, and fully compatible with downstream applications including Western blotting and mass spectrometry.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty