Would you like to log in again?
DURAN® Laboratory Bottles, without Screw Cap, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
218018609
490015-782EA 530.19 USD InStock
490015-782
DURAN® Laboratory Bottles, without Screw Cap, DWK Life Sciences
Bottles Glass Bottles
DURAN® laboratory glass bottles helps in storage, sample preparation, transport and Autoclaving media.
  • Standardised GL-thread and matching cap systems
  • Chemical resistance and shock resistance
  • Stable and easy to label
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty