Would you like to log in again?
100-Place Microscope Slide Box, White
Supplier: Heathrow Scientific
HS15995R
490015-822CS 1241.19 USD InStock
490015-822
100-Place Microscope Slide Box, White
Microscope Slide Boxes
Slide boxes are perfect for storage or transportation of standard size slides (75×25 mm and 3×1").
Molded, rigid boxes securely hold slides in grooved slots. Keep boxes safely on the benchtop during use or stack boxes for long-term storage. Boxes are secured with a rust-resistant nickel-plated clasp and hinge pin, and feature foam lining. Each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty