Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

81-Well Microtube Storage Boxes

Supplier: Heathrow Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

120036 120035 120039 120038 120037
490015-862BX 81.44 USD InStock
490015-862 490015-864 490015-866 490016-350 490015-868
81-Well Microtube Storage Boxes
Boxes Cryoboxes
Made of translucent, vibrantly colored polypropylene which allows for visibility inside the box and color coding identification. Tube capacity is indicated on the outside the box within a distinctive wave design. Quick visual identification with uniform wells and alphanumeric location identification along with a frosted labeling area.

  • Easy open and secure lid fitted with friction fit design
  • Stackable, saving valuable space in the lab
  • Can be used in temperatures as low as –80 °C
  • Autoclavable

Featuring molded grid lines on the lid, imprinted coordinates on the base, and a molded reference point on the lid.

Holds 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty