Would you like to log in again?
CappWash™ Microplate Washer, APS BE
Supplier: CAPP
W-12KIT-115V
490015-894EA 4536.59 USD InStock
490015-894
CappWash™ Microplate Washer, APS BE
Microplate Washers
CappWash™ is the ideal washing tool for small scale immuno assays
  • Compact and made for small-scale work
  • Unique two-in-one design
  • Simple washing; matches all well types
  • No programming and no electrical wiring connection required
The vacuum pump is available for 115V connections.
Fitting into even the tightest budget, CappWash™ is a perfect alternative for washing micro plates and strips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty