Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Infinite® M200 PRO Multimode Microplate Reader, Tecan
Supplier: Tecan
LGE140213-20BL
490016-280EA 44505 USD InStock
490016-280
Infinite® M200 PRO Multimode Microplate Reader, Tecan
Microplate Readers
The Infinite M200 provides a full range of leading detection methods. The Quad4 Monochromators of the Infinite M200 PRO provides exceptional sensitivity, and allows to select any wavelength from UV to NIR, and to perform absorbance, excitation and emission scans.
  • Fluorescence intensity top reading including TRF, with automated z-adjustment
  • Enhanced fluorescence intensity bottom reading with OR (Optimal Read) function
  • Extended emission wavelength ranges from 330 to 600 nm to 280 to 850 nm
It offers several detection modes for sample measurements in 6 to 384 well plates, PCR plates or cuvettes. The Infinite M200 PRO can be used for a wide range of biological assays and measurements including Protein, DNA/RNA quantification, ion channel and flux studies, ELISA and various assays.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty