Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ricca Chemical Buffer, pH 4.00

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

1501-16
490016-288EA 42.67 USD InStock
490016-288
Ricca Chemical Buffer, pH 4.00
Buffers pH Calibration Buffers
pH Buffer Reference Standard Solution for Calibration, pH 4.00 0.01 at 25°C.

  • Red Color Coded
  • Packaging: 500 mL

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty