Would you like to log in again?
MBP 100 µl Gel Loading Pipette Tips, 204 Tip Rack, Thermo Scientific
Supplier: Thermo Scientific
F010-R204S-100
490016-306BX 293.87 USD InStock
490016-306
MBP 100 µl Gel Loading Pipette Tips, 204 Tip Rack, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. First-Class Filter Tips: Specially designed for PCR work, these pipette tips prevent airborne contaminants from coming in contact with the barrel of the pipettor. Gel-loading Tips: Useful for a wide variety of sample loading applications such as protein separations, nucleic acid sequencing, and other gel loading applications. For use with but not limited to: Pipetman®, Biohit Proline®, Costar® Onepette, Eppendorf®, Finnpipette®, Jencons Sealpette®, Labpette™, MLA®, Nichiryo, Oxford-Benchmate®, Socorex®, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request. Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty