Would you like to log in again?
GeneMate Western Blot Boxes, Blotting Containers, MTC Bio
Supplier: MTC Bio
B1200-5 B1200-7 B1200-17 B1200-9 B1200-13 B1206 B1200-11 B1200-3 B1200-15BK B1200-7BK B1200-15 B1200-19
490016-636CS 1076.85 USD InStock
490016-636 490016-648 490016-634 490016-646 490016-632 490016-644 490016-652 490016-642 490016-650 490016-640 490019-528 490016-638
GeneMate Western Blot Boxes, Blotting Containers, MTC Bio
Blotting Apparatus
These western blot boxes are made of polystyrene and are ideal for halves, wholes, strips or minis.
  • Reduce antibody usage
  • Features 90° angle corners
These boxes feature easy-to-open hinged lid which reduces evaporation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty