Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Advansta Incubation Trays
Supplier: Advansta
L-07038-005 L-07036-003 L-07035-004 L-07034-005 L-07041-002 L-07037-002 L-07040-003 L-07039-004 L-07033-005 L-07031-005
490005-028PK 88.22 USD InStock
490005-028 490016-788 490016-800 490016-786 490016-798 490016-796 490016-794 490016-792 490016-790 490005-032
Advansta Incubation Trays
Trays/Dishes/Pots Laboratory Trays/Dishes
These incubation trays are convenient and easy to use for staining and washing gels and membranes.
  • Smooth interior- Protects membranes and gels from scratches
  • Lidded- To protect experiments from dust or debries
  • Non-binding surfaces prevent antibody loss
  • Convenient- Multiple sizes available for different applications
These lidded incubation trays have a perfectly smooth interior surface, making them ideal for staining and washing electrophoresis gels and membranes. Trays are manufactured to prevent adsorption of antibodies. Three sizes are available to hold different volumes or gel sizes, so you can find the right tray for your application.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty