Would you like to log in again?
Multilingual Wide Mouth Wash Bottles
Supplier: Dynalon
506495-0005
490017-202CS 171.69 USD InStock
490017-202
Multilingual Wide Mouth Wash Bottles
Bottles Wash Bottles
Multi-lingual wide mouth wash bottles feature a patented DripLok® vapor venting valve that prevents solvent drips.
  • Indelibly printed in four languages: English, French, German, and Spanish
  • Closure features precision jet-tip aperture for fine control.
Choose from four solvent labels, all including the new Globally Harmonized Symbols, NFPA diamond, risk phrases, and safety information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty