Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Screw Cap Tubes, 5.0 ml, MTC Bio

Supplier: MTC Bio

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

C2530 C2570 C2581 C2540-OB C2540
490019-562PK 222.4 USD InStock
490019-562 490019-792 490019-566 490019-568 490019-560
Screw Cap Tubes, 5.0 ml, MTC Bio
Tubes Test Tubes
These 5.0 ml tubes take all of the conveniences of standard 1.5 ml tubes and scales them up for larger volume and throughput.

  • Made of maximum clarity
  • Low-binding virgin polypropylene
  • Certified Rnase- and Dnase-free
  • Sterile tubes are also pyrogen free

The 16 mm tube diameter is identical to the diameter of standard 15 ml tubes, which ensures further compatibility with centrifuge rotors, more. In many applications, these tubes have replaced 15 ml tubes, dramatically reducing plastic waste, storage space, and shipping costs. The potential contamination of pipettor shafts is greatly reduced because the 56 mm total tube length (vs. 120 mm for standard 15 ml tubes) is short enough for standard 1 ml or 5 ml tips to reach the tube's conical bottom.

These screw cap tubes are supplied with 20 mm Ext.Ø caps that seal smoothly and completely with a single 360° turn. Useful from –86 to 110 °C, they are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from nonsterile bulk packaging with separate caps, or gamma sterilized rack packaging. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require approximately 60% less storage space.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty