Would you like to log in again?
MACROTUBE SNAP-CAP 5ML NON STERILE
Supplier: GeneMate
Macrotube^ VWR* Macrotube, 5 ml Series, Snap Cap, all colors, 16 x 56 mm, dual graduations, polypropylene, DNase/RNase free
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
MACROTUBE SNAP-CAP 5ML NON STERILE 490019-790 Pack of 200 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty